Order Entry
Finland
ContactUsLinkComponent
Conductor bottle
  765-0189
undefined
Conductor bottle
  765-0189
  • Description:
    Conductor bottle
  • Pk:
    1
For storage of electric charge. Consists of a glass cylinder with metal foil and a conductor ball on a rod.
  • Capacitance: about 1000 pF
  • Dimensions: 260 × 80 mm (diam)