Order Entry
Finland
Orders LinkContactUsLinkComponent
Plate capacitor
Plate capacitor
Plate capacitor
Catalog # 765-1189
Marketsource
Supplier:   PHYWE
Plate capacitor
Catalog # 765-1189
Supplier:  PHYWE
Marketsource
Plate capacitor
Plate capacitor
Plate capacitor
Price
2 870,00 €
Log in for availability
1 * 1 Each
Product Details & Documents
Precision capacitor for electrostatic tests e.g. examination of the relationship between charge, voltage and capacitance. Measurement of the dielectric constant and the electrostatic induction constant. The permanently mounted plate is well insulated while the movable plate is mounted directly in the stand. The distance between the capacitor plates can be adjusted with an accuracy of 0.1 mm. Distance between plates 0 to 70 mm.
  • Plate diameter 260 mm
  • Plate thickness 6 mm
Specifications
  • Description:
    Plate capacitor