Power supply
Power supply
Catalog # 765-0213
Marketsource
Supplier: CORNELSEN EXPERIMENTA
Jump to:
Product Details & Documents
Adjustable in three steps 3, 4, 5 V DC / 1,5 A. Equipped with 4 mm banana plugs and overload protection. Impact resistant plastic housing with mains plug.
Dimensions: 130×80×50 mm
Specifications
- Description:Power supply