Order Entry
Finland
ContactUsLinkComponent
 
2-Octyne ≥99.0% (by GC)
  TCIAO0125-5ML
 :  TCI
 :  
undefined
2-Octyne ≥99.0% (by GC)
  TCIAO0125-5ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 99.0 %

 

 

 :
 :110.2 g/mol
 :137°C
 :0,7596 g/cm³ (20 °C)
 :18°C
 :220-553-3
 :3295
 :3,II
 :GHS02
 :GHS08
 :2809-67-8
 :C₈H₁₄
 :MFCD00027302