Order Entry
Finland
ContactUsLinkComponent
3-(4-Aminophenyl)propionic acid ≥98.0% (by GC, titration analysis)
3-(4-Aminophenyl)propionic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
3-(4-Aminophenyl)propionic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light red to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    131.0 to 135.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together