5-Ethylphenazinium ethyl sulphate ≥95%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:10510-77-7
:
Formula:C₁₆H₁₈N₂O₄S
:334.4 g/mol
:M-Clarity™ quality level = MQ200
EINECS:234-044-9
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance (Colour):Yellow to Orange to Brown
- Appearance (Form):Powder
- Solubility (Colour):Yellow to Dark Yellow and Orange to Dark Orange and Yellow-Orange and Yellow-Brown and Orange-Brown and Dark Brown-Yellow
- Solubility (Turbidity)
50 mg/ml, EtOH:Clear - Infrared spectrum:Conforms to Structure
- Carbon:54.6 - 60.2 %
- Nitrogen:8.0 - 8.7 %
- Purity (HPLC):≥95.0 %
- Recommended Retest Period:3 years