Order Entry
Finland
ContactUsLinkComponent
EDTA dipotassium salt dihydrate
EDTA dipotassium salt dihydrate
Catalog # MOLE44541922-500G
Supplier:  Molekula
CAS Number:  
EDTA dipotassium salt dihydrate
Catalog # MOLE44541922-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :404.46 g/mol
Merck Index:13,03546
Melting Pt:272 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:217-895-0
CAS Number:25102-12-9
Formula:C₁₀H₁₄K₂N₂O₈·2H₂O
MDL Number:MFCD00150036