Order Entry
Finland
Orders LinkContactUsLinkComponent
(2,4-Dichlorophenoxy)acetic acid

(2,4-Dichlorophenoxy)acetic acid

Plastic bottle for solids, 500 g
Tuotenumero MOLEM63113303
Corrosive
Irritant
Marketsource
Toimittaja:   Molekula
(2,4-Dichlorophenoxy)acetic acid
Tuotenumero MOLEM63113303
Toimittaja:  Molekula
Marketsource
CAS numero:  94-75-7
(2,4-Dichlorophenoxy)acetic acid

Plastic bottle for solids, 500 g

CAS numero:  94-75-7
Price
77,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  2,4-D
  • CAS Number:94-75-7
  • Formula:(C₆H₃Cl₂)OCH₂COOH
  • MDL Number:MFCD00004300
  • Molecular Weight:221.04 g/mol
  • Merck Index:13,02825
  • Boiling Pt:346 °C (1013 hPa)
  • Melting Pt:137…139 °C
  • Density:1.563 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:202-361-1
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Cat. No.:
    MOLEM63113303

Specifications