Order Entry
Finland
ContactUsLinkComponent
1,1,3,3-Tetramethyl-2-thiourea for synthesis, Sigma-Aldrich®
Irritant
1,1,3,3-Tetramethyl-2-thiourea for synthesis, Sigma-Aldrich®
Catalog # 8.41854.9999
Supplier:  Merck
CAS Number:  
Irritant
1,1,3,3-Tetramethyl-2-thiourea for synthesis, Sigma-Aldrich®
Catalog # 8.41854.9999
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Solubility:
    32 g/l (20 °C)
  • Melting Point:
    75 - 78 °C
  • Molar Mass:
    132.22 g/mol
  • Boiling Point:
    245 °C (1013 hPa)
  • Vapor Pressure:
    1.9 hPa (25 °C)
  • Flash Point:
    100 °C

Specification Test Results

About this item

 :
 :132.23 g/mol
Boiling Pt:245 °C (1013 hPa)
Melting Pt:74…76 °C
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:220-488-0
 :GHS07
CAS Number:2782-91-4
Formula:C₅H₁₂N₂S
MDL Number:MFCD00008324