Order Entry
Finland
Orders LinkContactUsLinkComponent
3-(2-Hydroxyethyl)indole

3-(2-Hydroxyethyl)indole

Supplier:   Merck
3-(2-Hydroxyethyl)indole
Supplier:  Merck
Certificates
3-(2-Hydroxyethyl)indole

2 Options Available Below

Product Details & Documents
Synonyms:  3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol
  • CAS Number:526-55-6
  • Formula:C₁₀H₁₁NO
  • MDL Number:MFCD00005659
  • Molecular Weight:161.2 g/mol
  • Merck Index:12,09930
  • Boiling Pt:358 °C (1013 hPa)
  • Melting Pt:55…57 °C
  • EINECS:208-393-2

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g