
Diethylene glycol monobutyl ether, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:112-34-5
:
Formula:CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
MDL Number:MFCD00002881
:162.23 g/mol
Merck Index:13,01556
Boiling Pt:230.4 °C (1013 hPa)
Melting Pt:–68 °C
Density:0.954 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:100 °C
EINECS:203-961-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-68 °C
- Molar Mass:162.23 g/mol
- Boiling Point:226 - 234 °C (1013 hPa)
- Vapor Pressure:0.1 hPa (20 °C)
- Flash Point:105 °C
- Refractive Index:1.4258 (27 °C, 589 nm)
- Explosion Limit:0.7 - 5.3 %(V)
- Density:0.95 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
- Ignition Point:225 °C
- Viscosity kinematic:6.2 mm2/s (20 °C)
- Evaporation number:1200