Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium hydroxide
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol
FFCHFF9660500
:
FF CHEMICALS
:
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol
FFCHFF9660500
:
FF CHEMICALS
:
Pack type:
Glass bottle
Pk:
500 mL
Seq:
2725
:
:
56.11 g/mol
:
Ambient
:
215-181-3
:
3286
:
3,II
:
GHS02
:
GHS06
:
GHS05
:
GHS08
:
1310-58-3
:
KOH
:
MFCD00003553