N-Ethylmaleimide (NEM), Calbiochem®
Catalog # 34115-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Merck Index:13,03855
Boiling Pt:210 °C (1013 hPa)
Melting Pt:46 °C
ADR:6.1,II
:125.13 g/mol
EINECS:204-892-4
UN:2928
:GHS06
:GHS05
CAS Number:128-53-0
Formula:C₆H₇NO₂
MDL Number:MFCD00005509