Safranine O, Certifiable Grade
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:477-73-6
:
Formula:C₂₀H₁₉ClN₄
MDL Number:MFCD00011759
:350.85 g/mol
Density:1.447 g/cm³ (22 °C)
Storage Temperature:Ambient
EINECS:207-518-8
Specification Test Results
- Absorbance maximum:527 – 533 nm
- Dye Content (Anhydrous):≥80%
- Loss on Drying:≤4%
- P-15/P+15:1.1 – 1.32