
1-Butyl-3-methylimidazoliumhexafluorophosphate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:174501-64-5
:
Formula:C₈H₁₅F₆N₂P
MDL Number:MFCD03093295
:284.18 g/mol
Merck Index:14,1581
Boiling Pt:>350 °C
Melting Pt:8…10 °C
Density:1.365
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together