Order Entry
Finland
Orders LinkContactUsLinkComponent
3-(2-Hydroxyethyl)indole ≥97%
3-(2-Hydroxyethyl)indole ≥97%
3-(2-Hydroxyethyl)indole ≥97%
3-(2-Hydroxyethyl)indole ≥97%
Purity: 97% CAS No.: 526-55-6 Molecular Formula: C10H11NO Formula Weight: 161.20 Flash Point: 255°(491°F) Melting Point: 56-59° Boiling Point: 174°/2mm Beilstein Reference: 125553 Merck Reference: 14,9798 MDL No.: MFCD00005659
 :  3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol
  •  :526-55-6
  •  :C₁₀H₁₁NO
  •  :MFCD00005659
  •  :161.2 g/mol
  •  :14,9798
  •  :174 °C (2 mmHg)
  •  :56…59 °C
  •  :255 °C (491 °F)
  •  :Ambient
  •  :208-393-2
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    1 g
    1 * 1 g
    1 * 1 g - 51,80 €
    51,80 €
    1 * 1 g
       51,80 €
  • Glass bottle for solids
    5 g
    1 * 5 g
    1 * 5 g - 151,00 €
    151,00 €
    1 * 5 g
       151,00 €