Order Entry
Finland
ContactUsLinkComponent
 
Human Serum Albumin (HSA) Solution (25%), 50 ml
Catalog # GEMB800-120
undefined
Human Serum Albumin (HSA) Solution (25%), 50 ml
Catalog # GEMB800-120

Specifications

  • Serum format:
    Frozen
  • Sterility:
    Sterile
  • Description:
    Human serum albumin solution
  • Shipping conditions:
    Ships on dry ice
  • Storage temperature:
    Store frozen at −20...−10 °C
  • Pack type:
    Poly bottle
  • Pk:
    50 mL

Specifications

About this item

This diagnostic-grade product is an aqueous solution containing 25% human albumin (w/v).

It is prepared by cold-alcohol fractionation from pooled human plasma obtained from venous blood. The USP Pharmacopeia requires that all fractionators add sodium caprylate and sodium tryptophanate as a preservative.

Delivery: Ships on dry ice. Store frozen at −20 to −10 °C.
Caution: For cell culture, further manufacturing and research use only. Not for direct therapeutic use.