0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Ethyl propiolate
Ethyl propiolate ≥99%
Ethyl propiolate ≥99%
:
Thermo Fisher Scientific
Ethyl propiolate ≥99%
:
Thermo Fisher Scientific
:
623-47-2
:
:
C₅H₆O₂
:
MFCD00009184
:
118…120 °C
:
9 °C
:
0.963
:
23 °C (73 °F)
:
210-795-8
:
3,III
:
98.1 g/mol
:
3272
:
GHS02
:
GHS07
msds_ALFAA15068
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years