
Adipoyl dichloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-50-2
:
Formula:C₆H₈Cl₂O₂
MDL Number:MFCD00000759
:183.03 g/mol
Boiling Pt:105…107 °C (2 mmHg)
Density:1.255
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:203-876-4
UN:3265
ADR:8,II
:GHS05