
Pyridinium chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:628-13-7
:
Formula:C₅H₆ClN
MDL Number:MFCD00012802
:115.56 g/mol
Boiling Pt:222…224 °C
Melting Pt:∼ 145 °C
Density:1.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-027-4
:GHS07