
Methyl-2,5-dihydroxybenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2150-46-1
:
Formula:C₈H₈O₄
MDL Number:MFCD00016464
:168.15 g/mol
Melting Pt:86…87 °C
EINECS:218-427-8
:GHS07
Storage Temperature:Ambient