Safranine O 85%, pure, certified
:477-73-6
:
:C₂₀H₁₉ClN₄
:MFCD00011759
:350.85 g/mol
:1,447 g/cm³ (22 °C)
:207-518-8
:GHS05
:Ambient
- Appearance:Dark green to dark brown Powder
- Infrared spectrum:Conforms
- Dye content:≥80 %
- UV Lambda max:530 to 534 nm
- Absorption Ratio P-15/P+15:1.1 to 1.32
- BSC Certification:Passes