
Ammonium fluoride 98+% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12125-01-8
:
Formula:NH₄F
MDL Number:MFCD00011423
:37.04 g/mol
Merck Index:13,00524
Melting Pt:160 °C
Density:1,0088 to 1.0096 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:235-185-9
UN:2505
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White Adhering crystalline powder
- Sulfated ash:≤0.01 %
- Titration excess NaOH:≥98.0 %
- Chloride (Cl):≤0.001 %
- Sulfate (SO4):≤0.005 %
- Hexafluorosilicate (SiF6):≤0.1 %
- Iron (Fe):≤0.0005 %
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 % (in water)
- (NH4)HF2:≤1.0 %