
1-Butyl-3-methylimidazoliumhexafluorophosphate 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:174501-64-5
:
Formula:C₈H₁₅F₆N₂P
MDL Number:MFCD03093295
:284.18 g/mol
Boiling Pt:>340 °C (760 mmHg)
Melting Pt:6.5 °C
Density:1.3727 g/cm³
Flash Pt:≥ 110 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Viscous liquid
- Infrared spectrum:Conforms
- Water:≤1.0 % (K.F.)
- HPLC:≥98.0 %
Specification Test Results
Frequently Bought Together