Phenyl chloroformate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1885-14-9
:
Formula:C₇H₅ClO₂
MDL Number:MFCD00000637
Boiling Pt:188…189 °C (1013mbar)
Melting Pt:–28 °C
Density:1.244 g/cm³
Flash Pt:80 °C
Storage Temperature:Refrigerator
EINECS:217-547-8
ADR:6.1,II
:156.57 g/mol
UN:2746
:GHS06
:GHS05
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.5100 to 1.5120 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤50 APHA