9-Anthracenemethanol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1468-95-7
:
Formula:C₁₅H₁₂O
MDL Number:MFCD00001264
:208.26 g/mol
Boiling Pt:371 °C (1013 hPa)
Melting Pt:160…164 °C
Density:1,3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-998-5
ADR:6.1,III
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:160 to 164 °C
- GC:≥97.5 %