Safranine O 95%, pure, high purity biological stain
:477-73-6
:
:C₂₀H₁₉ClN₄
:MFCD00011759
:350.85 g/mol
:1,447 g/cm³ (22 °C)
:207-518-8
:GHS05
:Ambient
- Appearance:Green to red-brown Powder
- Infrared spectrum:Conforms
- Loss on drying:≤10 %
- Dye content:≥94 %