
3-(4-Hydroxyphenyl)propionic acid 98%
:501-97-3
:
:C₉H₁₀O₃
:MFCD00002778
:166.18 g/mol
:208 - 210 °C (14 -rr)
:126…130 °C
:1,336 g/cm³ (20 °C)
:Ambient
:207-931-3
:GHS07
- Appearance:Off-white to beige Powder or granules
- Infrared spectrum:Conforms
- Melting point:126 to 130 °C
- GC:≥97.5 %
- Titration with NaOH:≥97.5 %