
4,6-Dihydroxypyrimidine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1193-24-4
:
Formula:C₄H₄N₂O₂
MDL Number:MFCD00016733
:112.09 g/mol
Melting Pt:>300 °C
EINECS:214-772-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to orange Powder
- Infrared spectrum:Conforms
- Titration excess NaOH:≥97.5 %
- HPLC:≥97.5 %