Order Entry
Finland
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

Supplier:   GRATNELL'S
Storage Boxes
Supplier:  GRATNELL'S
Certificates
Storage Boxes

16 Options Available Below

Product Details & Documents

Polypropylene storage trays. The material is fire-resistant and resistant to most chemicals in the school environment. Stackable.

  • BSI tested and passed for heavy educational use
  • All trays fit standard school furniture
  • Lids and inserts also available
  • Fully recyclable
Product Family Options

Shallow tray F/1 (75 mm height)

Product Information

  • Colour
    W×D×H
    Availability
    Price
  • Trays
    Green
    312×427×75 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 25,00 €
    25,00 €
    1 * 1 Items
       25,00 €

  • Trays
    Red
    312×427×75 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 25,00 €
    25,00 €
    1 * 1 Items
       25,00 €

  • Trays
    Blue
    312×427×75 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 24,70 €
    24,70 €
    1 * 1 Items
       24,70 €

  • Trays
    Yellow
    312×427×75 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 21,30 €
    21,30 €
    1 * 1 Items
       21,30 €

Deep tray F/2 (150 mm height)

Product Information

  • Colour
    W×D×H
    Availability
    Price
  • Trays
    Translucent
    312×427×150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 42,50 €
    42,50 €
    1 * 1 Items
       42,50 €

  • Trays
    Green
    312×427×150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 35,00 €
    35,00 €
    1 * 1 Items
       35,00 €

  • Trays
    Red
    312×427×150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 35,00 €
    35,00 €
    1 * 1 Items
       35,00 €

  • Trays
    Blue
    312×427×150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 35,00 €
    35,00 €
    1 * 1 Items
       35,00 €

  • Trays
    Yellow
    312×427×150 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 30,70 €
    30,70 €
    1 * 1 Items
       30,70 €

Storage boxes F/3 (300 mm height)

Product Information

  • Colour
    W×D×H
    Availability
    Price
  • Trays
    Translucent
    312×430×300 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 53,30 €
    53,30 €
    1 * 1 Items
       53,30 €

  • Trays
    Green
    312×430×300 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 50,20 €
    50,20 €
    1 * 1 Items
       50,20 €

  • Trays
    Red
    312×430×300 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 53,30 €
    53,30 €
    1 * 1 Items
       53,30 €

  • Trays
    Blue
    312×430×300 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 49,90 €
    49,90 €
    1 * 1 Items
       49,90 €

  • Trays
    Yellow
    312×430×300 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 53,30 €
    53,30 €
    1 * 1 Items
       53,30 €

Lids and tray carriers

Product Information

  • Colour
    W×D×H
    Availability
    Price
  • 765-0964
    Tray carrier with handle
    Tray carrier with handle
    - mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items

Runners

Product Information

  • Colour
    W×D×H
    Availability
    Price
  • 763-0316
    Wood furniture runners
    Wood furniture runners
    - mm
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - 8,60 €
    8,60 €
    1 * 2 Items
       8,60 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.