Order Entry
Finland
Orders LinkContactUsLinkComponent
Cell building
Cell building
Cell structure model, SOMSO-Plast®
Catalog # 763-0452
Supplier:   SOMMER MARCUS
Cell building
Catalog # 763-0452
Supplier:  SOMMER MARCUS
Cell building
Cell building
Cell structure model, SOMSO-Plast®
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
The 10000x magnification model shows the structure of an animal cell. Cell plasma with mitochondria, centrioles, endoplasmic reticulum with and without ribosomes. Cell nucleus with nuclear membranes, pores, nucleols, chromatin.
Delivery information: 22 cm high cell model mounted on a stand with a base.
Specifications
  • Description:
    Cell structure model
  • Material:
    SOMSO-Plast®