Order Entry
Finland
ContactUsLinkComponent
Platinum loop with glass handle
Platinum loop with glass handle
Catalog # 765-1396
Supplier:  LABGLAS
undefined
Platinum loop with glass handle
Catalog # 765-1396
Supplier:  LABGLAS
Specifications
  • Description:
    Platinum loop with glass handle
  • Pk:
    1
Specifications
About this item
Total length approx. 200 mm, glass shaft 150×5 mm. Thread diameter 0,5 mm, inner diameter of the loop 1,5 mm.