Order Entry
Finland
ContactUsLinkComponent
Dicotyledon plant stem
Dicotyledon plant stem
Catalog # 763-0138
Supplier:  3B Scientific
undefined
Dicotyledon plant stem
Catalog # 763-0138
Supplier:  3B Scientific
Specifications
  • Weight:
    1,2 kg
  • Description:
    Dicotyledonous plant stem, Phaseolus vulgaris
  • W×D×H:
    340×340×260 mm
  • Pk:
    1
Specifications
About this item
Phaseolus vulgaris. Model of dicotyledonous stem (beans) magnified 250X.
Stem cell types are shown longitudinally and in cross section.