Order Entry
Finland
Orders LinkContactUsLinkComponent
Laser pointer

Laser pointer

Laser pointer, pen model, without ball pen, 14×14×130 mm
Catalog # 765-1014
Supplier:   KVANT LIFE SCIENCE
Laser pointer
Catalog # 765-1014
Supplier:  KVANT LIFE SCIENCE
Laser pointer
Laser pointer

Laser pointer, pen model, without ball pen, 14×14×130 mm

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

Power: 1 mW, Laser class II.

  • Wavelength: 645 nm
  • Operating time: about 6 hours
Specifications
  • Description:
    Laser pointer, pen model, without ball pen
  • W×D×H:
    14×14×130 mm