Order Entry
Finland
Orders LinkContactUsLinkComponent
(R)-(-)-1-Phenyl-1,2-ethanediol

(R)-(-)-1-Phenyl-1,2-ethanediol

Glass bottle for solids, 1 g
Catalog # MOLE36796198-1G
Marketsource
Supplier:   Molekula
(R)-(-)-1-Phenyl-1,2-ethanediol
Catalog # MOLE36796198-1G
Supplier:  Molekula
Marketsource
CAS Number:  16355-00-3
(R)-(-)-1-Phenyl-1,2-ethanediol

Glass bottle for solids, 1 g

CAS Number:  16355-00-3
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (R)-1-Phenylethane-1,2-diol, (R)-(-)-α,β-Dihydroxyethylbenzene, (R)-(-)-Phenylethane-1,2-diol, (R)-(-)-Phenyl-1,2-ethanediol
  • CAS Number:16355-00-3
  • Formula:C₈H₁₀O₂
  • MDL Number:MFCD00064262
  • Molecular Weight:138.17 g/mol
  • Boiling Pt:272 °C (1013 hPa)
  • Melting Pt:67…69 °C
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.