Order Entry
Finland
ContactUsLinkComponent
 
Sodium-L(+)-glutamate monohydrate
Catalog # MOLE40359230-1KG
Supplier:  Molekula
CAS Number:  
Sodium-L(+)-glutamate monohydrate
Catalog # MOLE40359230-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :187.13 g/mol
Merck Index:13,06278
Melting Pt:225…240 °C
Flash Pt:>225 °C
Storage Temperature:Ambient
EINECS:240-313-1
CAS Number:6106-04-3
Formula:C₅H₈NNaO₄·H₂O
MDL Number:MFCD00150138