0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
4-Bromo-3-chloro-7-methoxyquinoline
4-Bromo-3-chloro-7-methoxyquinoline 95%
4-Bromo-3-chloro-7-methoxyquinoline 95%
Irritant
4-Bromo-3-chloro-7-methoxyquinoline 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Bromo-3-chloro-7-methoxyquinoline 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1208452-88-3
:
Formula:
C₁₀H₇BrClNO
MDL Number:
MFCD13619692
:
272.53 g/mol
:
GHS07