Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Phenazine-1,6-diol
Phenazine-1,6-diol
Phenazine-1,6-diol
Irritant
Phenazine-1,6-diol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Phenazine-1,6-diol
Supplier:
Apollo Scientific
About this item
CAS Number:
69-48-7
:
Formula:
C₁₂H₈N₂O₂
MDL Number:
MFCD01708477
:
212.20 g/mol
:
GHS07