Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tetraphenylphosphonium bromide
Tetraphenylphosphonium bromide 98%
Tetraphenylphosphonium bromide 98%
Irritant
Tetraphenylphosphonium bromide 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Tetraphenylphosphonium bromide 98%
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
2751-90-8
:
Formula:
C₂₄H₂₀BrP
MDL Number:
MFCD00011915
Melting Pt:
294…296 °C
:
419.3 g/mol
:
GHS07
Related Information
Documentation
Certificates
Search by Lot Number
Documentation