Supelco® Organic Reference Standard, Pyridine solution
Pyridine solution, 2000 μg/ml in methanol, Certified reference material, 1
Catalog # 48305-U
Supplier: Merck
Jump to:
Product Details & Documents
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311
Specifications
Specifications
- Concentration:2000 μg/ml in methanol
- Pk:1
- Standards:Certified reference material
- Description:Pyridine solution
- Cat. No.:48305-U
- M-Clarity quality level:MQ100
- Storage conditions:2...30 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Flash point:11 °C
- CAS No.:110-86-1
- Shipping conditions:STD



