Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Chlorobenzene-[D5] 2,000 µg/ml in methanol, Supelco®
Chlorobenzene-[D5] 2,000 µg/ml in methanol, Supelco®
Chlorobenzene-[D5] 2,000 µg/ml in methanol, Supelco®
Catalog # 48086.
Supplier:
Merck
CAS Number:
Expand image
Chlorobenzene-[D5] 2,000 µg/ml in methanol, Supelco®
Catalog # 48086.
Supplier:
Merck
CAS Number:
Specifications
Pk:
1
CAS No.:
3114-55-4
Description:
Chlorobenzene-[D5] 2,000 µg/ml in methanol
Formula:
C₆ClD₅
Description:
Chlorobenzene-[D5]
Specifications
About this item
Product Details
Product Details