Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-OG488 acid
6-OG488 acid ≥95% [equivalent to Oregon Green® 488 carboxylic acid, 6-isomer]
6-OG488 acid ≥95% [equivalent to Oregon Green® 488 carboxylic acid, 6-isomer]
6-OG488 acid ≥95% [equivalent to Oregon Green® 488 carboxylic acid, 6-isomer]
Catalog # AATB712
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
6-OG488 acid ≥95% [equivalent to Oregon Green® 488 carboxylic acid, 6-isomer]
Catalog # AATB712
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
25 mg
Seq:
2315
Specifications
About this item
Product Details
:
CAS Number:
195136-53-9
Product Details