
7-Hydroxy-4-methylcoumarin-3-acetic acid ≥95%
Catalog # AATB554
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:234.21 g/mol
ADR:5,II
:GHS07
CAS Number:5852-10-8
Formula:C₁₂H₁₀O₅
MDL Number:MFCD00467589