Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fluo-5F AM
Fluo-5F AM Cell permeant
Fluo-5F AM Cell permeant
AATB20560
:
AAT BIOQUEST
:
Fluo-5F AM Cell permeant
AATB20560
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
500 µG
Seq:
1418
:
:
1100.91 g/mol
:
5,II
:
GHS07