0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Fluo-2 potassium salt
Fluo-2 potassium salt
Fluo-2 potassium salt
Fluo-2 potassium salt
AATB20493
:
AAT BIOQUEST
:
Fluo-2 potassium salt
AATB20493
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
500 µG
Seq:
1418
:
:
891.1 g/mol
:
5,II
:
GHS07