Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Acridine orange
Acridine orange 10 mg/ml ≥95% in water
Acridine orange 10 mg/ml ≥95% in water
Acridine orange 10 mg/ml ≥95% in water
Catalog # AATB17503
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Acridine orange 10 mg/ml ≥95% in water
Catalog # AATB17503
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
10 mL
Seq:
2275
Specifications
About this item
Product Details
:
:
265.35 g/mol
CAS Number:
494-38-2
Formula:
C₁₇H₁₉N₃
MDL Number:
MFCD00005029
Product Details