Order Entry
Finland
ContactUsLinkComponent
Cyanine 3.5 maleimide ≥90% [equivalent to Cy3.5® Maleimide]
Cyanine 3.5 maleimide ≥90% [equivalent to Cy3.5® Maleimide]
  AATB149
 :  
undefined
Cyanine 3.5 maleimide ≥90% [equivalent to Cy3.5® Maleimide]
  AATB149
 :  

 

  • Pk:
    1 mg
  • Seq:
    2070

 

 

 :
 :1198.24 g/mol