0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Cyanine 3.5 acid
Cyanine 3.5 acid ≥95% [equivalent to Cy3.5® acid]
Cyanine 3.5 acid ≥95% [equivalent to Cy3.5® acid]
Catalog # AATB147
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Cyanine 3.5 acid ≥95% [equivalent to Cy3.5® acid]
Catalog # AATB147
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
1005.04 g/mol
Product Details