HRP-Biotin Conjugate
Catalog # AATB11022
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:5 mg
- Description:HRP-biotin conjugate
Specifications
About this item
Biotinylated horse radish peroxidase (HRP) is an excellent tool for biological detections with ELISA and other enzyme-coupled assays.
- Highly useful antibody label
- Widely used for immunoassays
- Multiple applications
Caution: For research use only.