Order Entry
Finland
ContactUsLinkComponent
Helix Fluor™ 545 Succinimidyl Ester
Helix Fluor™ 545 Succinimidyl Ester
Catalog # AATB250
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Helix Fluor™ 545 Succinimidyl Ester
Catalog # AATB250
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Description:
    Helix Fluor™ 555 succinimidyl ester
  • Purity:
    90%

Specifications

About this item

FAM, TET, VIC, JOE, NED, TAMRA and ROX fluorescent dyes are frequently used for labelling oligos.

  • Multiple Bioconjugation Applications
  • Excellent Quality
Caution: For research use only.